| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600007862.mm.1 |
NR_040389.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007862.mm.1 |
NR_040389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007862.mm.1 |
TC1600001009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600007864.mm.1 |
NR_040389.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007864.mm.1 |
NR_040389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007864.mm.1 |
TC1600001009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600007865.mm.1 |
NR_040389.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007865.mm.1 |
NR_040389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600007865.mm.1 |
TC1600001009.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1442574_PM_at |
- |
- |
| Mouse Expression Set 430 |
1442574_at |
- |
- |