Assay Details
Gene Symbol: Gm38424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038023.1 | - | 3 - 4 | 1651 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112338 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971696 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164423 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717731 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15346 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123511 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05523308 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123511 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05523318 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04407340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04407336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04642156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04407341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04252777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578919 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04355181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04927181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04407337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04407344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04956308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04407347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04355182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04927175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04755303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23578944 | - | - |
