Assay Details
Gene Symbol: Gm7854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028417.1 | 7 - 8 | 975 | 79 | ||
GenBank mRNA | AK041797.1 | - | 7 - 8 | 976 | 79 | |
AK138349.1 | - | 5 - 6 | 421 | 79 | ||
AV378858.2 | - | 2 - 3 | 221 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254654 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624006 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805255 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508029 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833535 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914895 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20613 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132774 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905574 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942019 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324827 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203073 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670148 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695553 | NM_053251.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580368 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887251 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266672 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505130 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779087 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788162 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823631 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968339 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904017 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136510 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714654 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419724 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564846 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973650 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244861 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473128 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76224 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18737 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127615 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147550 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29375 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110399 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101979 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05550304 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01085882 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110399 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05550333 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05550368 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101979 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05550396 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101979 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05550368 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04249688 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04246599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04141147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23644307 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1457085_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10529799 | - | - |
Mouse Genome 430 2.0 Array | 1457085_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457085_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 665934_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 665934_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500021259.mm.1 | TC0500002311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500021259.mm.1 | NR_028417.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500021259.mm.1 | NR_028417.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500021260.mm.1 | TC0500002311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500021260.mm.1 | NR_028417.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500021260.mm.1 | NR_028417.1 | Refseq |
Mouse Expression Set 430 | 1457085_at | - | - |
Mouse Expression Set 430 | 1457085_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 377736 | - | - |
