Assay Details
| Gene Symbol: Cdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019753.4 | NP_062727.1 | 10 - 11 | 1428 | 149 | |
| GenBank mRNA | AF177669.1 | - | 10 - 11 | 1324 | 149 | |
| AK089791.1 | - | 10 - 11 | 1395 | 150 | ||
| AK155633.1 | - | 10 - 11 | 1429 | 149 | ||
| AK171733.1 | - | 10 - 11 | 1351 | 149 | ||
| BC042891.1 | - | 10 - 11 | 1346 | 4208360 | 149 | |
| D87912.1 | - | 10 - 11 | 1373 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh17
Genomic Map