Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400000648.mm.1 |
NM_019753.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000648.mm.1 |
NM_019753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000648.mm.1 |
TC0400000093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000647.mm.1 |
NM_019753.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000647.mm.1 |
NM_019753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000647.mm.1 |
TC0400000093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000650.mm.1 |
NM_019753.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000650.mm.1 |
NM_019753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000650.mm.1 |
TC0400000093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000649.mm.1 |
NM_019753.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000649.mm.1 |
NM_019753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000649.mm.1 |
TC0400000093.mm.1 |
MTA TC |