Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300004628.mm.1 |
NM_001083318.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004628.mm.1 |
NM_001286844.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004628.mm.1 |
NM_012051.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004628.mm.1 |
TC0300000697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300004618.mm.1 |
NM_001083318.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004618.mm.1 |
NM_001286844.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004618.mm.1 |
NM_012051.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004618.mm.1 |
TC0300000697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300004623.mm.1 |
NM_001286844.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004623.mm.1 |
TC0300000697.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300004626.mm.1 |
NM_001083318.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004626.mm.1 |
NM_001286844.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004626.mm.1 |
NM_012051.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004626.mm.1 |
TC0300000697.mm.1 |
MTA TC |