Assay Details
Gene Symbol: Etv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083318.2 | NP_001076787.1 | 3 - 4 | 378 | 127 | |
NM_001286844.1 | NP_001273773.1 | 3 - 4 | 694 | 127 | ||
NM_012051.4 | NP_036181.3 | 3 - 4 | 375 | 127 | ||
GenBank mRNA | AF156530.1 | - | 2 - 3 | 304 | 127 | |
AF218539.2 | - | 3 - 4 | 361 | 127 | ||
AK089640.1 | - | 3 - 4 | 325 | 127 | ||
AK147552.1 | - | 1 - 2 | 2501 | 127 | ||
AK155957.1 | - | 3 - 4 | 316 | 127 | ||
AK156824.1 | - | 3 - 4 | 323 | 127 | ||
AK163509.1 | - | 3 - 4 | 343 | 127 | ||
BC057151.1 | - | 3 - 4 | 312 | 127 | ||
BC078636.1 | - | 3 - 4 | 312 | 4918332 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
