Assay Details
| Gene Symbol: Pgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175013.2 | NP_778178.3 | 5 - 6 | 940 | 77 | |
| GenBank mRNA | AK035507.1 | - | 5 - 6 | 940 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Pgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175013.2 | NP_778178.3 | 5 - 6 | 940 | 77 | |
| GenBank mRNA | AK035507.1 | - | 5 - 6 | 940 | 77 | |
Set Membership: |
|