Assay Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019971.2 | NP_064355.1 | 5 - 6 | 1945 | 79 | |
| GenBank mRNA | AF117608.1 | - | 5 - 6 | 1121 | 79 | |
| AF266467.1 | - | 5 - 6 | 1945 | 79 | ||
| AF286725.1 | - | 5 - 6 | 924 | 79 | ||
| AK033734.1 | - | 5 - 6 | 1674 | 79 | ||
| AK042767.1 | - | 5 - 6 | 1692 | 79 | ||
| AK052947.1 | - | 5 - 6 | 1197 | 79 | ||
| BC029099.1 | - | 6 - 7 | 1248 | 79 | ||
| BC037696.1 | - | 5 - 6 | 1132 | 4008749 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfc
Genomic Map