Assay Details
| Gene Symbol: Ldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077398.2 | NP_001070866.1 | 2 - 3 | 492 | 136 | |
| NM_001286348.1 | NP_001273277.1 | 2 - 3 | 492 | 136 | ||
| NM_010698.4 | NP_034828.3 | 2 - 3 | 492 | 136 | ||
| GenBank mRNA | AB221652.1 | - | 2 - 3 | 239 | 136 | |
| AK146860.1 | - | 2 - 3 | 426 | 136 | ||
| AK147160.1 | - | 2 - 3 | 402 | 136 | ||
| BC079611.1 | - | 2 - 3 | 385 | 30360903 | 136 | |
| CN670130.1 | - | 2 - 3 | 454 | 136 | ||
| U89487.1 | - | 2 - 3 | 381 | 136 | ||
| U89489.1 | - | 2 - 3 | 381 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldb2
Genomic Map