Assay Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027088.2 | NP_081364.1 | 6 - 7 | 570 | 70 | |
| GenBank mRNA | AB047820.1 | - | 6 - 7 | 471 | 70 | |
| AK079374.1 | - | 6 - 7 | 586 | 70 | ||
| AK129108.1 | - | 6 - 7 | 478 | 70 | ||
| AK170576.1 | - | 6 - 7 | 571 | 70 | ||
| AK171561.1 | - | 6 - 7 | 572 | 70 | ||
| BC042489.1 | - | 6 - 7 | 521 | 70 | ||
| BC050901.1 | - | 6 - 7 | 533 | 6402443 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bap1
Genomic Map