Assay Details
| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019975.3 | NP_064359.2 | 5 - 6 | 477 | 107 | |
| GenBank mRNA | AJ132139.1 | - | 5 - 6 | 475 | 107 | |
| AK005505.1 | - | 5 - 6 | 464 | 107 | ||
| AK041686.1 | - | 5 - 6 | 473 | 107 | ||
| AK050078.1 | - | 5 - 6 | 477 | 107 | ||
| AK161488.1 | - | 457 | 107 | |||
| BC021360.1 | - | 5 - 6 | 408 | 5011451 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hacl1
Genomic Map