Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400002886.mm.1 |
NM_030166.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002886.mm.1 |
NM_030166.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002886.mm.1 |
TC1400000336.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002887.mm.1 |
NM_030166.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002887.mm.1 |
NM_030166.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002887.mm.1 |
TC1400000336.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002888.mm.1 |
NM_030166.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002888.mm.1 |
NM_030166.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002888.mm.1 |
TC1400000336.mm.1 |
MTA TC |