Assay Details
| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028083.4 | NP_082359.1 | 1 - 2 | 80 | 64 | |
| GenBank mRNA | AK011243.1 | - | 1 - 2 | 81 | 64 | |
| AK038378.1 | - | 1 - 2 | 50 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011243.1, NM_028083.4 transcripts.
Set Membership: |
|

Chaf1b
Genomic Map