Assay Details
| Gene Symbol: S100a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011313.2 | NP_035443.1 | 1 - 2 | 338 | 127 | |
| GenBank mRNA | BM932500.1 | - | 1 - 2 | 350 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011313.2 transcripts.
Set Membership: |
|

S100a6
Genomic Map