Assay Details
| Gene Symbol: Mark2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080388.2 | NP_001073857.1 | 5 - 6 | 1039 | 84 | |
| NM_001080389.2 | NP_001073858.1 | 5 - 6 | 1039 | 84 | ||
| NM_001080390.2 | NP_001073859.1 | 5 - 6 | 756 | 84 | ||
| NM_007928.3 | NP_031954.2 | 5 - 6 | 1039 | 84 | ||
| GenBank mRNA | AK045329.1 | - | 5 - 6 | 613 | 84 | |
| AK154864.1 | - | 5 - 6 | 756 | 84 | ||
| AK172444.1 | - | 5 - 6 | 999 | 84 | ||
| AK172664.1 | - | 5 - 6 | 682 | 84 | ||
| AK220191.1 | - | 6 - 7 | 605 | 84 | ||
| BC058556.1 | - | 5 - 6 | 976 | 5289799 | 84 | |
| X70764.1 | - | 5 - 6 | 493 | 84 | ||
More Information
Set Membership: |
|

Mark2
Genomic Map