Assay Details
| Gene Symbol: Mark2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080388.2 | NP_001073857.1 | 7 - 8 | 1176 | 98 | |
| NM_001080389.2 | NP_001073858.1 | 7 - 8 | 1176 | 98 | ||
| NM_001080390.2 | NP_001073859.1 | 7 - 8 | 893 | 98 | ||
| NM_007928.3 | NP_031954.2 | 7 - 8 | 1176 | 98 | ||
| GenBank mRNA | AK045329.1 | - | 7 - 8 | 750 | 98 | |
| AK172444.1 | - | 7 - 8 | 1136 | 98 | ||
| AK172664.1 | - | 7 - 8 | 819 | 98 | ||
| AK215655.1 | - | 3 - 4 | 187 | 98 | ||
| AK217185.1 | - | 2 - 3 | 136 | 98 | ||
| AK220191.1 | - | 8 - 9 | 742 | 98 | ||
| BC058556.1 | - | 7 - 8 | 1113 | 5289799 | 98 | |
| X70764.1 | - | 7 - 8 | 630 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mark2
Genomic Map