Assay Details
| Gene Symbol: Etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025794.2 | NP_080070.2 | 3 - 4 | 619 | 78 | |
| GenBank mRNA | AK002483.1 | - | 3 - 4 | 572 | 78 | |
| AK037969.1 | - | 3 - 4 | 536 | 78 | ||
| AK075673.1 | - | 3 - 4 | 570 | 78 | ||
| AK085493.1 | - | 3 - 4 | 588 | 78 | ||
| AK141684.1 | - | 3 - 4 | 573 | 78 | ||
| AK152624.1 | - | 3 - 4 | 588 | 78 | ||
| AK169686.1 | - | 3 - 4 | 524 | 78 | ||
| BC012522.1 | - | 3 - 4 | 568 | 2648103 | 78 | |
| BC057670.1 | - | 3 - 4 | 586 | 2810976 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etfdh
Genomic Map