Assay Details
| Gene Symbol: Ncapg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019438.1 | NP_062311.1 | 7 - 8 | 1218 | 78 | |
| GenBank mRNA | AJ237585.1 | - | 2 - 3 | 281 | 78 | |
| AK017756.1 | - | 7 - 8 | 1112 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncapg
Genomic Map