Assay Details
| Gene Symbol: Rbm46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277170.1 | NP_001264099.1 | 3 - 4 | 807 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P580634 | NM_001146328.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P580634 | NM_001146328.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK004611.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK030613.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK033107.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK054329.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK075829.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK163696.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK165587.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK178472.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK207349.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | AK210410.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | BC005541.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | NM_001039533.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346965 | NM_053181.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111609 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P580634 | NM_001146328.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P580634 | NM_001146328.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P580634 | NM_001146328.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P580634 | NM_001146328.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10498976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498975 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498974 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498972 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300016739.mm.1 | NM_001277170.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016739.mm.1 | TC0300002228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300016740.mm.1 | NM_001277170.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016740.mm.1 | TC0300002228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300016743.mm.1 | NM_001277170.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016743.mm.1 | TC0300002228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300016742.mm.1 | NM_001277170.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016742.mm.1 | TC0300002228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300016744.mm.1 | NM_001277170.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016744.mm.1 | TC0300002228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300016737.mm.1 | NM_001277170.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300016737.mm.1 | TC0300002228.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 795870 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252250 | - | - |

Rbm46
Genomic Map