Assay Details
| Gene Symbol: Gucy1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161796.1 | NP_001155268.1 | 9 - 10 | 1313 | 68 | |
| NM_017469.4 | NP_059497.1 | 9 - 10 | 1313 | 68 | ||
| GenBank mRNA | AF020339.1 | - | 9 - 10 | 1287 | 68 | |
| AF297083.1 | - | 9 - 10 | 1287 | 68 | ||
| AK139406.1 | - | 9 - 10 | 1276 | 68 | ||
| AK147128.1 | - | 9 - 10 | 1248 | 68 | ||
| BC050945.1 | - | 9 - 10 | 1291 | 6822142 | 68 | |
| BC056995.1 | - | 9 - 10 | 1291 | 6822142 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy1b3
Genomic Map