Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300004173.mm.1 |
NM_021370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004173.mm.1 |
NM_021370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004173.mm.1 |
TC0300000636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300004170.mm.1 |
NM_021370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004170.mm.1 |
NM_021370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004170.mm.1 |
TC0300000636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300004174.mm.1 |
NM_021370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004174.mm.1 |
NM_021370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300004174.mm.1 |
TC0300000636.mm.1 |
MTA TC |