Assay Details
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175360.2 | NP_780569.1 | 3 - 4 | 482 | 79 | |
GenBank mRNA | AK075574.1 | - | 3 - 4 | 482 | 79 | |
AK135228.1 | - | 3 - 4 | 500 | 79 | ||
AK146929.1 | - | 3 - 4 | 560 | 79 | ||
AK164386.1 | - | 3 - 4 | 492 | 79 | ||
AK166348.1 | - | 3 - 4 | 556 | 79 | ||
BC029548.1 | - | 3 - 4 | 522 | 5012064 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
