Assay Details
| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011481.3 | NP_035611.3 | 6 - 7 | 1568 | 78 | |
| GenBank mRNA | AK049582.1 | - | 6 - 7 | 829 | 78 | |
| BC120633.1 | - | 6 - 7 | 1547 | 40129556 | 78 | |
| BC125325.1 | - | 6 - 7 | 1544 | 40129840 | 78 | |
| D26186.1 | - | 6 - 7 | 1414 | 78 | ||
| D49427.1 | - | 6 - 7 | 1582 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srms
Genomic Map