Assay Details
| Gene Symbol: Eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007906.2 | NP_031932.1 | 5 - 6 | 1140 | 82 | |
| GenBank mRNA | BC018235.1 | - | 5 - 6 | 1140 | 4504385 | 82 |
| L26479.1 | - | 5 - 6 | 906 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eef1a2
Genomic Map