Assay Details
| Gene Symbol: Slc4a1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009206.2 | NP_033232.2 | 3 - 4 | 1007 | 104 | |
| GenBank mRNA | AF035526.1 | - | 4 - 5 | 890 | 104 | |
| AK138398.1 | - | 3 - 4 | 962 | 104 | ||
| BC072588.1 | - | 3 - 4 | 990 | 104 | ||
| BC090964.1 | - | 3 - 4 | 559 | 30625511 | 104 | |
More Information
Set Membership: |
|

Slc4a1ap
Genomic Map