Assay Details
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147201.2 | NP_671734.1 | 4 - 5 | 539 | 138 | |
GenBank mRNA | AF302138.1 | - | 4 - 5 | 538 | 138 | |
AK046142.1 | - | 4 - 5 | 533 | 138 | ||
AK077440.1 | - | 4 - 5 | 575 | 138 | ||
AK170501.1 | - | 4 - 5 | 555 | 138 | ||
BC004756.1 | - | 4 - 5 | 509 | 3154089 | 138 | |
BC018463.1 | - | 4 - 5 | 513 | 3598438 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
