Assay Details
Gene Symbol: Agbl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048192.2 | NP_001041657.1 | 8 - 9 | 1578 | 81 | |
NM_001294262.1 | NP_001281191.1 | 7 - 8 | 1606 | 81 | ||
NM_174849.3 | NP_777274.1 | 7 - 8 | 1495 | 81 | ||
NR_122116.1 | 7 - 8 | 1562 | 81 | |||
NR_122117.1 | 7 - 8 | 1562 | 81 | |||
GenBank mRNA | AK029676.1 | - | 7 - 8 | 1496 | 81 | |
AK034894.1 | - | 7 - 8 | 1584 | 81 | ||
AK035680.1 | - | 5 - 6 | 1063 | 81 | ||
AK083656.1 | - | 7 - 8 | 1445 | 81 | ||
BC057349.1 | - | 7 - 8 | 1496 | 6813901 | 81 | |
DQ867034.1 | - | 7 - 8 | 1491 | 81 | ||
DQ867035.1 | - | 7 - 8 | 1491 | 81 | ||
DQ867036.1 | - | 8 - 9 | 1578 | 81 | ||
DQ867037.1 | - | 7 - 8 | 1491 | 81 | ||
DQ867038.1 | - | 7 - 8 | 1491 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
