Assay Details
Gene Symbol: Rarres1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164763.1 | NP_001158235.1 | 1 - 2 | 365 | 128 | |
GenBank mRNA | AK077393.1 | - | 1 - 2 | 365 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
