Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146207.2 | NP_666319.2 | 2 - 3 | 295 | 98 | |
| GenBank mRNA | AK144802.1 | - | 2 - 3 | 277 | 98 | |
| AK150237.1 | - | 2 - 3 | 274 | 98 | ||
| AK166251.1 | - | 2 - 3 | 277 | 98 | ||
| AK168215.1 | - | 2 - 3 | 277 | 98 | ||
| AK170722.1 | - | 2 - 3 | 277 | 98 | ||
| AK182330.1 | - | 1 - 2 | 55 | 98 | ||
| AK213794.1 | - | 1 - 2 | 55 | 98 | ||
| AK217224.1 | - | 1 - 2 | 55 | 98 | ||
| BC010211.1 | - | 2 - 3 | 273 | 3969515 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cul4a
Genomic Map