Assay Details
| Gene Symbol: Ripk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023663.6 | NP_076152.2 | 3 - 4 | 670 | 64 | |
| GenBank mRNA | AF302127.3 | - | 3 - 4 | 673 | 64 | |
| AK077233.1 | - | 3 - 4 | 855 | 64 | ||
| AK145203.1 | - | 3 - 4 | 652 | 64 | ||
| BC057871.1 | - | 3 - 4 | 633 | 5294197 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ripk4
Genomic Map