Assay Details
| Gene Symbol: Antxrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172808.2 | NP_766396.2 | 5 - 6 | 706 | 69 | |
| GenBank mRNA | AK077206.1 | - | 5 - 6 | 707 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Antxrl
Genomic Map