Assay Details
| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008978.2 | NP_033004.1 | 8 - 9 | 822 | 129 | |
| GenBank mRNA | AK017003.1 | - | 8 - 9 | 885 | 129 | |
| AK029493.1 | - | 8 - 9 | 820 | 129 | ||
| D64141.1 | - | 7 - 8 | 821 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn20
Genomic Map