Assay Details
| Gene Symbol: Bst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009763.3 | NP_033893.2 | 3 - 4 | 464 | 77 | |
| GenBank mRNA | AK041059.1 | - | 3 - 4 | 443 | 77 | |
| AK164028.1 | - | 1 - 2 | 244 | 77 | ||
| BC033422.1 | - | 3 - 4 | 439 | 77 | ||
| D31788.1 | - | 3 - 4 | 443 | 77 | ||
| L32812.1 | - | 3 - 4 | 447 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bst1
Genomic Map