Assay Details
| Gene Symbol: Sdr16c6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080710.2 | NP_001074179.1 | 4 - 5 | 669 | 79 | |
| GenBank mRNA | AK076465.1 | - | 4 - 5 | 650 | 80 | |
| BC125450.1 | - | 4 - 5 | 669 | 40129965 | 79 | |
| BC125452.1 | - | 4 - 5 | 669 | 40129967 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sdr16c6
Genomic Map