Assay Details
| Gene Symbol: Ercc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081221.1 | NP_001074690.1 | 16 - 17 | 3004 | 75 | |
| GenBank mRNA | BC132447.1 | - | 12 - 13 | 1943 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ercc6
Genomic Map