Assay Details
| Gene Symbol: Ercc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081221.1 | NP_001074690.1 | 5 - 6 | 1474 | 89 | |
| GenBank mRNA | AK028576.1 | - | 5 - 6 | 1466 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ercc6
Genomic Map