Assay Details
Gene Symbol: AI464131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085515.2 | NP_001078984.1 | 1 - 2 | 78 | 69 | |
GenBank mRNA | AK173117.1 | - | 1 - 2 | 60 | 69 | |
BY246173.1 | - | 1 - 2 | 35 | 69 | ||
BY283520.1 | - | 1 - 2 | 78 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001085515.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
metabolic process
positive regulation of insulin-like growth factor receptor signaling pathway
skeletal muscle fiber development
positive regulation of protein kinase B signaling
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
protein binding
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity
carbohydrate metabolic process
metabolic process
positive regulation of insulin-like growth factor receptor signaling pathway
skeletal muscle fiber development
positive regulation of protein kinase B signaling
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
protein binding
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406557 | NM_001085515.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406557 | AK173117.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406557 | BC036141.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406557 | NM_001085515.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406557 | AK173117.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406557 | BC036141.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961311 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650036 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50537 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406557 | NM_001085515.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406557 | AK173117.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406557 | BC036141.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406557 | NM_001085515.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406557 | AK173117.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406557 | BC036141.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05212666 | - | - |
Whole Mouse Genome G4122A | A_51_P406557 | NM_001085515.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406557 | AK173117.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406557 | BC036141.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406557 | NM_001085515.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406557 | AK173117.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406557 | BC036141.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22862443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03610575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03867713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862427 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03867707 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04038998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03867705 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03775822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03628198 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03775832 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03204531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03746807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03481288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03775821 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03610570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03775839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03867710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03409047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799608 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22862474 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10512252 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512253 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435417_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10512251 | - | - |
Mouse Genome 430 2.0 Array | 1435417_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435417_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329828_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329828_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400019933.mm.1 | NM_001085515.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019933.mm.1 | NM_001085515.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019933.mm.1 | TC0400002469.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400019929.mm.1 | NM_001085515.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019929.mm.1 | NM_001085515.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019929.mm.1 | TC0400002469.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400019934.mm.1 | NM_001085515.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019934.mm.1 | NM_001085515.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019934.mm.1 | TC0400002469.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400019930.mm.1 | NM_001085515.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019930.mm.1 | NM_001085515.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019930.mm.1 | TC0400002469.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400019931.mm.1 | NM_001085515.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019931.mm.1 | NM_001085515.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400019931.mm.1 | TC0400002469.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435417_PM_at | - | - |
Mouse Expression Set 430 | 1435417_at | - | - |
Murine Genome U74v2 Set | 104034_PM_at | - | - |
Murine Genome U74v2 Set | 104034_at | - | - |
Mouse Genome 430A 2.0 Array | 1435417_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435417_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435417_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435417_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 545821 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 101500022 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2670472 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222435 | - | - |
