Assay Details
| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181325.4 | NP_851842.1 | 4 - 5 | 696 | 96 | |
| GenBank mRNA | AF133914.1 | - | 4 - 5 | 544 | 96 | |
| AK052758.1 | - | 4 - 5 | 628 | 96 | ||
| AK142223.1 | - | 4 - 5 | 566 | 96 | ||
| AK145966.1 | - | 4 - 5 | 566 | 96 | ||
| AK154988.1 | - | 4 - 5 | 672 | 96 | ||
| BC061143.1 | - | 4 - 5 | 337 | 30276598 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061143.1 transcripts.
Set Membership: |
|

Slc25a15
Genomic Map