Assay Details
| Gene Symbol: 4930431F12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073013.1 | 6 - 7 | 921 | 81 | ||
| GenBank mRNA | AK015267.1 | - | 6 - 7 | 922 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LIM domain binding 2 | Ldb2 |
AI035351, AW146358, CLIM1, CLP-36, Ldb3 |
-172.3kb | Anti-sense | Chr. 5 - 44472131 - 44799760 on NCBI Build GRCm38 |
| Downstream |
DNA segment, Chr 5, ERATO Doi 615, expressed |
D5Ertd615e | 4930435H24Rik | 137.2kb | Anti-sense | Chr. 5 - 45109944 - 45295343 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1400001966.rn
Set Membership: |
|

4930431F12Rik
Genomic Map