Assay Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177111.3 | NP_796085.3 | 3 - 4 | 126 | 134 | |
| GenBank mRNA | AK170006.1 | - | 3 - 4 | 174 | 134 | |
| BC070471.1 | - | 3 - 4 | 126 | 30626499 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc66
Genomic Map