Assay Details
| Gene Symbol: Ppp2r3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163415.1 | NP_001156887.1 | 1460 | 58 | ||
| NM_011154.2 | NP_035284.2 | 1226 | 58 | |||
| NR_028110.2 | 1599 | 58 | ||||
| NR_102716.1 | 1717 | 58 | ||||
| GenBank mRNA | AF050165.1 | - | 1223 | 58 | ||
| AK054122.1 | - | 1599 | 58 | |||
| AK143212.1 | - | 1503 | 58 | |||
| BC027415.1 | - | 1203 | 4235453 | 58 | ||
| BC059852.1 | - | 1491 | 6402777 | 58 | ||
| BC095938.1 | - | 381 | 58 | |||
| BC096544.1 | - | 906 | 6834351 | 58 | ||
More Information
Set Membership: |
|

Ppp2r3d
Genomic Map