Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400001093.mm.1 |
NM_010281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001093.mm.1 |
NM_010281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001093.mm.1 |
TC0400000169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001090.mm.1 |
NM_010281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001090.mm.1 |
NM_010281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001090.mm.1 |
TC0400000169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001091.mm.1 |
NM_010281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001091.mm.1 |
NM_010281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001091.mm.1 |
TC0400000169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001092.mm.1 |
NM_010281.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001092.mm.1 |
NM_010281.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001092.mm.1 |
TC0400000169.mm.1 |
MTA TC |