Assay Details
| Gene Symbol: Atp6v0d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175406.3 | NP_780615.2 | 6 - 7 | 888 | 123 | |
| GenBank mRNA | AB088358.1 | - | 6 - 7 | 844 | 123 | |
| AK075745.1 | - | 6 - 7 | 928 | 123 | ||
| AK089507.1 | - | 6 - 7 | 813 | 123 | ||
| AK137586.1 | - | 6 - 7 | 870 | 123 | ||
| AK170072.1 | - | 6 - 7 | 862 | 123 | ||
| AK170405.1 | - | 6 - 7 | 861 | 123 | ||
| AK170578.1 | - | 6 - 7 | 832 | 123 | ||
| AK170659.1 | - | 6 - 7 | 837 | 123 | ||
| AY145896.1 | - | 6 - 7 | 888 | 123 | ||
| AY517482.1 | - | 6 - 7 | 880 | 123 | ||
| BC087899.1 | - | 6 - 7 | 822 | 30504725 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0d2
Genomic Map