Assay Details
Gene Symbol: Best1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011913.2 | NP_036043.2 | 9 - 10 | 1314 | 76 | |
GenBank mRNA | AY450427.1 | - | 8 - 9 | 1138 | 76 | |
BC139060.1 | - | 8 - 9 | 1129 | 8862082 | 76 | |
BC139061.1 | - | 8 - 9 | 1129 | 8862083 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
