Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017083.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017085.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017082.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017084.mm.1 |
TC0300002273.mm.1 |
MTA TC |