Assay Details
| Gene Symbol: Sh3d19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D89677.2 | - | 2 - 3 | 66 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D89677.2 transcripts.
Set Membership: |
|

Sh3d19
Genomic Map