Assay Details
Gene Symbol: 4930556G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040655.1 | 5 - 6 | 624 | 92 | ||
GenBank mRNA | AK016143.1 | - | 5 - 6 | 625 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Eph receptor A7 | Epha7 |
Cek11, Ebk, Ehk3, Hek11, Mdk1 |
-1.9mb | Sense | Chr. 4 - 28813044 - 28967511 on NCBI Build GRCm38 |
Downstream | mitogen-activated protein kinase kinase kinase 7 | Map3k7 |
B430101B05, C87327, Tak1 |
1.1mb | Sense | Chr. 4 - 31962296 - 32023470 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
