Assay Details
Gene Symbol: Fhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021318.3 | NP_067293.1 | 2 - 3 | 339 | 107 | |
GenBank mRNA | AF083394.1 | - | 2 - 3 | 287 | 107 | |
AK006414.1 | - | 2 - 3 | 328 | 107 | ||
BC048473.1 | - | 2 - 3 | 339 | 6770515 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
