Assay Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019992.3 | NP_064376.1 | 4 - 5 | 374 | 106 | |
| GenBank mRNA | AB036058.1 | - | 4 - 5 | 364 | 106 | |
| AK041474.1 | - | 4 - 5 | 375 | 106 | ||
| AK153661.1 | - | 4 - 5 | 453 | 106 | ||
| AK162655.1 | - | 4 - 5 | 456 | 106 | ||
| BC145828.1 | - | 4 - 5 | 389 | 40131131 | 106 | |
| BC145830.1 | - | 4 - 5 | 390 | 40131133 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stap1
Genomic Map